Safety-Coated Medium Round Bottles, Clear, Wide Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Reduce breakage and lab mishaps with these safety coated clear medium round bottles.
If the glass breaks, the plastisol coating contains the substance in the bottle long enough to allow for proper disposal.
Customers Who Bought This Also Bought