Nalgene® Disposable Polypropylene Robotic Reservoirs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene Disposable Polypropylene Robotic Reservoirs come with either a convoluted bottom design or a flat bottom design.
- Convoluted design minimizes dead volume and has baffles to reduce splashing
- Convoluted bottom design is most effective with a 96-well format
- Flat Bottom design can be used with single, 6-, 12-, 96-, 384- and 1536-well formats
These reservoirs are compatible with most robotic arms, plate stackers, plate cranes, and platform configurations.
Certifications: Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004).
Customers Who Bought This Also Bought