VWR® Talon® Clamp Replacement Sleeves
Fournisseur: VWR International
Certificats
A propos de cet article
Replacement sleeves are easily removed for cleaning or replacement.
- Vinyl or fiberglass fabrication
- Fiberglass sleeves suitable for high-temperature applications
- Easily removed for cleaning
Available in vinyl or fiberglass. Fiberglass sleeves are recommended for all applications above 100 °C (212 °F).
Customers Who Bought This Also Bought