ContactUsLinkComponent
Removable Needle Compression Fitting, 1/16 Inch, Hamilton Company
Removable Needle Compression Fitting, 1/16 Inch, Hamilton Company
# de catalogue 89187-002
Fournisseur:  Hamilton
Rn Compression Fitting 1/16in
Removable Needle Compression Fitting, 1/16 Inch, Hamilton Company
# de catalogue 89187-002
Fournisseur:  Hamilton
Spécifications
  • Description:
    Hamilton 1/16 Inch Removable Needle Compression Fitting
  • Cat. no.:
    89187-002
  • Maximum Pressure:
    100 psi
  • Supplier No.:
    55751-01
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
The 1/16 inch connector is designed to attach 1/16 inch PEEK tubing directly to a small hub removable needle (RN) connection. Tightening the RN nut compresses the conical PFA ferrule into the PEEK cup ferrule creating a seal between the syringe barrel and the PEEK tubing. The connection requires no modifications to the PEEK tubing. Range of syringe is 2.5 -100 uL.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Ordering information: This product consists of one large bore RN nut, five PFA ferrules, and five PEEK cup ferrules.
Customers Who Bought This Also Bought