ContactUsLinkComponent
VWR® Refrigerated Circulating Baths, 240 V
VWR® Refrigerated Circulating Baths, 240 V
Fournisseur:  VWR International
Certificats
VWR® Refrigerated Circulating Baths, 240 V
VWR® Refrigerated Circulating Baths, 240 V
Fournisseur:  VWR International
Certificats
Spécifications
  • Catalog No:
  • 89202-972
  • 89202-992
  • 89202-968
  • 89203-012
  • 89202-988
  • 89202-964
  • 89203-004
  • 89202-980
  • 89203-024
  • 89203-022
  • 89202-976
  • 89202-996
  • Max. suction flow rate:
  • 3.2 gpm (12.2 l/min.), except units with MX Controller
  • 3.2 gpm (12.2 l/min.), except units with MX Controller
  • 3.2 gpm (12.2 l/min.), except units with MX Controller
  • 3.2 gpm (12.2 l/min.), except units with MX Controller
  • 3.9 gpm (14.7 l/min.), except units with MX Controller
  • 3.2 gpm (12.2 l/min.), except units with MX Controller
Spécifications
Informations connexes