About this item
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40° to 200°C (depending on the controller).
- Offer precise temperature control
- Available in a wide range of sizes
- Three controller options
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large, LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Specifications
- Catalog No:
- 89203-014
- 89203-010
- 89203-006
- 89203-002
- 89202-998
- 89202-994
- 89202-990
- 89202-986
- 89202-982
- 89202-978
- 89202-974
- 89202-970
- 89202-966
- 89202-962
- 89202-958
- Max. suction flow rate:
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller
- 3.9 gpm (14.7 l/min.), except units with MX Controller