Mobile Stainless Steel Instrument Table, Lakeside Manufacturing
Fournisseur: Lakeside
Certificats
A propos de cet article
Stainless steel instrument table.
- All welded construction increases durability and makes surface smoother for cleaning
- 18 gauge reinformcements supply extra security for larger models
- Nesting tables save space
18 gauge stainless steel top (and shelf if provided) is easy to clean and sanitize. 1" square 16-gauge stainless steel legs for superior strength. 3" locking casters ensure table is stationary when needed.
Customers Who Bought This Also Bought