VWR® Immersion Circulators
# de catalogue 89202-886
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Max. pressure:1.8 psi
- Working temperature range:Ambient +10 °C to 135 °C (275°F)
- Working Depth:18.4 cm (7¼")
- Electrical:120V, 60Hz
- Overall dimensions:10.9L x 9.7W x 35.8H cm (49/32 x 313/16 x 143/32")
- Temperature stability:±0.07°C
- Max. pressure flow rate:12.8 l/min
Spécifications
A propos de cet article
Create your own circulating bath system using your own vessel or a VWR® open bath tank.
- User-adjustable temperature limits and alarms
- Convenient slide control
- Single-point calibration
Large, 8.3 cm (31/4") universal icon and display. User-adjustable high- and low-temperature limits and alarms, as well as integral pump and heater coil protection. Convenient slide control allows easy adjustments of flow rate. Single-point calibration.
Certifications: ETL listed. CE marked. Complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. cETLus compliant.
Ordering information: 240V/50Hz models are also available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought