384 Deep Well, Small Volume Polypropylene Microplates, Greiner Bio-One
Fournisseur: Greiner Bio-One
Certificats
A propos de cet article
Greiner Bio-One 384 Deep Well Small Volume Polypropylene Microplates are especially suited for direct compound transfer and preparation of assay-ready plates and pre-dilutions.
- Polypropylene construction
- RNase-, DNase-, and human DNA-free
- Nonpyrogenic
- Efficient use of space with small footprint size
- Unique well geometry minimizes dead volume
Also, these plates can be used as storage plates and the standardized footprint allows efficient integration with automated systems
Black and white versions are available upon request.
These plates are designed with a unique well geometry that allows liquid transfers with minimal dead volume, using pipet tips or pin tools. Low dead volumes conserve valuable compounds that are lost in conventional well designs.
Ordering information: Lid not included.
Customers Who Bought This Also Bought