Nalgene® Colored CryoBoxes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene® Colored CryoBoxes come in 5 grid colors for easy-to-identify inventory management, providing safe and economical storage of up to 81 cryogenic samples.
These colored boxes hold 1.0 to 2.0 ml cryogenic storage vials in a 9 x 9 array, and feature a usable temperature range of –196°C to +121°C. The colors match popular NALGENE cryogenic vial color coders.
The transparent lid allows easy viewing of contents and the lids have a numbered grid for quick sample identification.
Customers Who Bought This Also Bought