CApture™ Portable Fuming System, Labconco
Fournisseur: Labconco
Certificats
A propos de cet article
The CApture™ Portable Fuming System quickly fixes latent fingerprints at the crime scene using microburst or traditional CA fuming methods, while protecting the user from hazardous fumes
Includes programmable heater, fuming chamber, exhaust blower and filter, hook, self and filter.
Its patent-pending design combines all the components needed in one compact system: a heater to heat the Super Glue®; a shatter-resistant, clear protective chamber to contain the fumes as they react with the latent prints; an exhaust blower; and particulate/carbon exhaust filter. No ducting is required. Since prints may be easily developed at the scene, irrelevant evidence may be ruled out thus saving valuable laboratory processing time and storage space.
Customers Who Bought This Also Bought