A propos de cet article
Nalgene® Polyolefin labels remain unaffected by water, UV light or chemicals.
The labels are for use with all writing devices except pencil and formatted for easy setup in word processing programs, but are not intended for use in photocopiers or laser printers. They resist ultraviolet light, mildew, chemicals, and dirt, will not discolor, crack, or tear and adhere well to any plastic, glass, or metal surface.
Nalgene® Polyolefin labels remain unaffected by water, UV light or chemicals.
The labels are for use with all writing devices except pencil and formatted for easy setup in word processing programs, but are not intended for use in photocopiers or laser printers. They resist ultraviolet light, mildew, chemicals, and dirt, will not discolor, crack, or tear and adhere well to any plastic, glass, or metal surface.
These die-cut labels have pressure-sensitive adhesive backs with rounded corners that enable easy removal from backing paper.