Accessories for RapidVap® Vertex™ Dry Evaporator, Labconco
Fournisseur: Labconco
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
Sulfuric acid 2.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$119.71 to $1,363.18
Hydrochloric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Fournisseur: VWR International
$99.45 to $1,061.29
TURKEY RED BLOOD CELLS 10% Washed Pooled Cells, Rockland Immunochemicals
Fournisseur: Rockland Immunochemical
$597.24