VWR® Maximum High Tack Shoe Covers
Fournisseur: VWR International
Certificats
A propos de cet article
High Tack Shoe Covers are manufactured from specially formulated 100 gsm polyurethane material to create optimal traction and durability.
High Tack Shoe Covers are manufactured from specially formulated 100 gsm polyurethane material to create optimal traction and durability. These shoe covers are fully heat sealed to prevent any liquid penetration.
- High coefficient of friction
- 100-gram basis weight
- Heat-sealed seams
- Ideal for wet surfaces
These shoe covers are fully heat sealed to prevent any liquid penetration.
Certifications: UL Listed.
Spécifications
- Catalog No:
- 414005-084
- 414005-083
- Garment application:
- Laboratory
- Laboratory