EXP® Fittings for 10-32 Ports and ¹/₁₆" Stainless Steel Tubing
Fournisseur: Optimize
Certificats
A propos de cet article
The EXP Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between ¹/₃₂ or ¹/₁₆" tubing and any 6-32, 6-40 or 10-32 port.
- Rated to 20000+ psi (1400+ bar)
- Tested to 30000 psi
- Auto-adjusting ZDV Connection
- Intended for many repeat uses
- No more cut-off and replacement of swaged ferrules
- Simple and versatile - can be used in UHPLC and HPLC applications
- Use with EXP® Nuts and Holders
The patented titanium hybrid ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The hybrid ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.
Customers Who Bought This Also Bought