Nalgene® InVitro™ Biotainer Bottles, Sterile, PETG, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene® InVitro™ Biotainer Bottles are designed specifically for bio-pharmaceutical manufacturers.
The bottles are made from crystal clear PETG, feature a space-saving square shape with ribbed hand grips (except 125 ml), and the closures feature a polyethylene foam core liner.
They reduce the risk of carry-over contamination by eliminating the cost of cleaning, sterilizing and associated validation.
Certifications: Meet USP 87, 88. Lot tested for pyrogenicity.
Customers Who Bought This Also Bought