Nalgene® Unitary™ Safety Wash Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene® Unitary™ Safety Wash Bottles are molded in one piece which eliminates possible contamination from a delivery tube and offers safety from chemical splashes when removing the closure.
The bottles also feature a leakproof linerless polypropylene closure, and are red for easy identification.
Customers Who Bought This Also Bought