Nalgene® Economy Polymethylpentene Graduated Cylinders, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene® Economy Polymethylpentene Graduated Cylinders exhibit excellent chemical resistance and a generous pour spout for easily dispensing liquids.
These cylinders are translucent, break resisitant and non-wetting. They feature large rounded bases are ribbed for reinforcement and stability, easy-to-read molded in graduations that won't rub off, and no meniscus to confuse readings allow for accurate measurements. These cylinders are calibrated "To Contain" and "To Deliver" at 20°C, and feature good temperature resistance up to 135°C (275°F).
Specifications
- Catalog No:
- 24780-425
- 24780-403
- 24780-469
- 24780-447
- 24780-505
- 24780-480
- 24780-527
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)