Nalgene® Economy Polypropylene Graduated Cylinders, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Nalgene® Economy PP Graduated Cylinders exhibit excellent chemical resistance and have a generous pour spout for easily dispensing liquids
They have large rounded bases and are ribbed for reinforcement and stability.The cylinders are calibrated TC and TD at 20°C. Also they feature good temperature resistance up to 135°C.
Spécifications
- Catalog No:
- 24780-265
- 24780-287
- 24780-268
- 24780-323
- 24780-301
- 24780-367
- 24780-345
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall