A propos de cet article
Greiner Bio -One disposal bags are ideal for the disposal of contaminated laboratory items.
Also available upon request with imprint "biohazard."
- Autoclavable
- Polypropylene construction
- Disposal bags may be marked with 'biohazard' upon request
Standard bags are made of polypropylene foil for sterilization in the steam autoclave.
Also available upon request with imprint 'biohazard'.
Oxygen transport rate of 50 µm PP foil.