PCR Plate Sealers, Greiner Bio-One
Fournisseur: Greiner Bio-One
Certificats
A propos de cet article
Greiner Bio-One offers four different varieties of adhesive microplate sealing films and foils.
- Acrylate adhesive coating
- Transparency differs between models
- Different seals are ideal for different applications
EASYseal™ is a transparent sealer used for protection against contamination and evaporation
All plate sealers are coated with an acrylate adhesive. VIEWseal™ is coated with a pressure sensitive silicone adhesive.
SILVERseal™ is an aluminum sealer ideal for PCR applications and storage of sample material and active agents. VIEWseal™ is highly transparent and is most suited for protein crystallography. AMPLIseal™ is ideal for real-time PCR.
Customers Who Bought This Also Bought