Envoy® Overnight Ready Engineered Shippers, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
A simple, economical solution for less complex shipping needs.
1-Day Shippers are guaranteed for 30 hours. 2-Day Shippers are guaranteed for 54 hours.
- Up to 54 hours of protection
- Ideal for one to two day deliveries
- Easy to assemble
A simple, economical solution for less complex shipping needs.
1-Day Shippers offer 30 hours of protection
Perfect for last mile pharmacy shipments. Ideal for one- to two-day delivery; this series hold product between 2 and 8°C (36 and 46°F). Seasonal packouts are simple to follow and easy to assemble. Reusable gels are environmentally friendly.
2-Day Shippers protect for 54 hours.
Certifications: Compatible to ISTA-5.
Customers Who Bought This Also Bought