Advanced TC™ Treated 384-Well Microplate, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Advanced Tissue Culture (TC™) treated 384 Well Microplates, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
- Virgin polystyrene construction
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield.
Plates available in Small Volume™ LoBase design as the two 4-25 µL microplates.
Sterilized by radiation. All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
Ordering information: Lid included. Small Volume formatted microplates come with ultra low profile lid.
Frequently Bought Together