Advanced TC™ Treated 96-Well Cell Culture Microplate, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
Certificates
About this item
Advanced Tissue Culture (TC) treated 96 Well Microplates, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
- Sterile
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
- Noncytotoxic
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield.
All plates come with lid.
Ordering information: Lid included with Condensation Rings.
Customers Who Bought This Also Bought