RotoFlex Tube Rotator, Argos Technologies
Fournisseur: Argos Technologies
Certificats
A propos de cet article
The RotoFlex will accommodate a variety of tubes from 1.5-50 mL by simply changing the rotisserie
To change the rotation direction- simply touch the rotisserie paddle while in motion and the RotoFlex automatically begins to rotate in the opposite direction. RotoFlex can be used in incubators and cold room settings. Unit is supplied with base unit and rotisseries (1 pack of each size).
No tools are required. The mixing angle can be adjusted from 0-90° and each of the 4 tube panels can be aligned independently. The 360° rotation ensures thorough mixing of all samples at the same time while rotating at 22 rpm. Ideal applications include hybridization, DNA purification, sample mixing and resuspension, protein binding and immunoprecipitation.
Customers Who Bought This Also Bought