VACUUBRAND® VACUU•LAN® Mini-Network, BrandTech
Catalog # 89176-810
Supplier: Brandtech
Specifications
- Description:VACUUBRAND® VACUU•LAN® Mini-Network
- Dimensions:24x213/16x 51/2 (LxWxD)
- Environmentally Preferable:Energy Efficient
- Cat. no.:89176-810
- Supplier No.:2614455
Specifications
About this item
Increase the utility of your oil-free vacuum pump with the VACUUBRAND VACUULAN® Local Vacuum Network
- Three flow-controled ports from one vacuum source
- Check valves minimize interference
- Mounts to walls, benches or lab frames
- Flow path made of corrosion resistant materials
With the pre-assembled VACUULAN® Mini-Network, you can have three VACUULAN® ports that can be mounted to ring stands or lab frames. Connector rods can be removed for mounting to walls or benches. Ports include manual flow control for continuously variable pumping speed adjustment. Suitable for use with VACUUBRAND® or other PTFE diaphragm pumps for chemistry applications.
The network includes three vacuum ports than have integrated check valves to minimize interference between applications.
Customers Who Bought This Also Bought