Spécifications
- Description:BRAND HandyStep® Repeating Pipette
- Cat. no.:89176-652
- Supplier No.:705110
Spécifications
A propos de cet article
The BRAND HandyStep® S repeating pipette provides unsurpassed versatility in a purely mechanical repeating pipette.
Comfortable. The slim design of the HandyStep® S is lightweight, well-balanced, and provides ambidextrous one-handed volume selection and dispensing. A new Tip ejector provides contact-free tip removal.
Innovative polymer construction provides a dependable, corrosion resistant, robust instrument. The purely mechanical design means no batteries to charge or replace–a convenient integrated volume table provides a quick reference for dispensed volumes.
Included shelf/rack mount also attaches easily to BRAND Transferpette® S carousel stand.
In conjunction with BRAND PD-Tip™ syringe tips, the rugged HandyStep S offers nearly sixty different volume settings with accuracy and precision.