Micro Titer Luminescence Microplate Strips, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Micro Titer Luminescence polystyrene Microplate Strips have flat bottoms and are configured in breakable 1 x 12 well strips
The microlite 1 strips have a hydrophobic surface for a preferred binding of biomolecules with hydrophobic domains.
The microlite 2 strips have a slightly hydrophilic surface for increased binding of biomolecules with hydrohilic/hydrophobic character.
These strips can be broken down into any number of wells and placed in the frame, which provides a high degree of flexibility in assay design
Customers Who Bought This Also Bought