Sodium thiocyanate ≥98%, crystals ACS
# de catalogue 700002-050
Fournisseur: Spectrum Chemicals
Small Business Enterprise
Numéro CAS: 540-72-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass, amber
- Size:500 g
- Cat. no.:700002-050
- Supplier No.:S1495-500GM
Spécifications
Résultats des tests de spécification
- Assay:98% Min.
- Sodium thiocyanate ≥98%, crystals ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sodium rhodanide
Point de Fusion:287 °C
Densité:1,735 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 287 °C
:Pt Eclair:|Point eclaire:
:M.W. 81,07 g/mol
Index Merck:13,09401
:Thiocyanate de sodium
EINECS:208-754-4
:287
:GHS07
Température d'entreposage:Température ambiante
:12,09463
Densité:1.735 g/cm³ (20 °C)
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 287 °C
:Flash Pt:
:Flash Pt:
:M.W. 81.07 g/mol
:Sodium thiocyanate
Formule:NaSCN
MW:81.07 g/mol
Température d'entreposage:Ambient
Numéro MDL:MFCD00011123
Numéro CAS:540-72-7
Customers Who Bought This Also Bought