PYREXPLUS® Coated Aspirator Bottles with Bottom Sidearm, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PYREXPLUS®, coated.
These aspirator bottles have a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and is autoclavable at 121 °C and resistant to thermal shock. Bottles have a tubular sidearm outlet (10 mm Ø) near the bottom for attachment of flexible plastic tubing and are very useful as a delivery/storage container for solutions. The tubulation is not coated to allow easy connection of standard size tubing. These bottles feature sturdy ring necks with a heavy duty rims and use No. 2 rubber stoppers.
Warning: Do not place over direct heat or flame. Do not heat about 121 °C moist heat or 110 °C dry heat.
Spécifications
- Catalog No:
- 16331-392
- 16331-395
- 89091-680
- 89091-672
- 89091-690
- 89091-686
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey