ContactUsLinkComponent
ExiSpin
ExiSpin
# de catalogue 89170-878
Fournisseur:  Bioneer
Numéro CAS:  
ExiSpin
# de catalogue 89170-878
Fournisseur:  Bioneer
Numéro CAS:  
Spécifications
  • Size:
    Each
  • Enzyme name:
    ExiSpin
  • Cat. no.:
    89170-878
  • Supplier No.:
    A-7040
Spécifications
A propos de cet article
ExiSpin has the functions of a vortexer as well as a microcentrifuge and will significantly save you time in the lab
The time and strength of centrifugation/mixing is fully programmable. The numbers of spin-mix-spin cycles can be set (1-999). After this input, a single button automatically starts the process as follows:
It provides reproducible results using protocols optimized to samples by preventing the possibilities of experimental errors. Four 8-strips tubes can be handled simultaneously, which enable 32 PCR/QPCR mixture tubes ready to run at once. Twelve 1.5 ml tubes can be loaded for simultaneous cell lysis and subsequent centrifuge-down. The programmed speed and time quarantine the reproducibility of each application.
ExiSpin comes complete with rotors for 4 x 8-strip tubes and 12 x 1.5/2.0 ml tubes. Use it for Minipreps, to resuspend oligos, set up you PCR reactions, or any task that requires vortexing in micro test tube or 0.2 ml PCR tubes.
Customers Who Bought This Also Bought