Microtiter, 96-Well Deep Well Microplates, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Microtiter 96-well deep-well microplates are virgin-grade polypropylene and automation friendly
These microplates are chemical resistant and biologically inert and feature a temperature range of -196° to +121°C.
Customers Who Bought This Also Bought