Nalgene® Water Quality Membrane, Cellulose Nitrate, Gridded, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Membrane turns black when wet.
Membrane turns black when wet.
Membranes contain no extractables that will inhibit or stimulate growth of bacteria and have no significant effect on the pH of the medium. They exhibit neither toxicity nor chemical or physical changes induced by the sterilization technique.
Certifications: Certified to meet requirements of Standard Methods for water quality work.
Packaging: Sterile and individually wrapped.
Customers Who Bought This Also Bought