VWR® TBS Buffer, 20X Liquid
# de catalogue 97064-338
Fournisseur: VWR
Numéro CAS:
Spécifications
- Size:4 L
- Supplier No.:J640-4L
Spécifications
A propos de cet article
A 1X solution of Tris Buffered Saline (TBS) contains 140 mM Sodium Chloride, 3.0 mM Potassium Chloride and 25 mM Tris.
Customers Who Bought This Also Bought