HyLite® 47-402 Nitrile Gloves, Fully Coated, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
A versatile alternative to cotton, light leather, and PVC, this special nitrile formulation provides an excellent combination of flexibility, mechanical resistance and oil repellency.
- Silicone-free
- Comfortable hand shape
The natural hand shape is easier to wear for long periods.
Use in general industry, maintenance, janitorial services, sanitation, metal fabrication, oil processing and refining, petrochemical, public utilities, and logistics and warehousing industries. Uses include assembling and inspecting engine parts and small components; general handling; sheet and light metals handling; stamping operations; gas and electricity supply; refuse collection; and shipping and receiving.
Ensures cleanliness of metal sheets, important in the steel and motor industries.
Certifications: ANSI level 1 cut protection. ANSI level 4 abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.
Spécifications
- Catalog No:
- 80092-600
- 80092-598
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Knit
- Knit
- Finish:
- Fully Coated
- Fully Coated
- Cuff style:
- Knit Wrist
- Knit Wrist
- EN norm:
- EN 388 (3111)
- EN 388 (3111)