Jet Guard Safety Air Guns, Guardair
# de catalogue 300052-508
Fournisseur: Safety & Industrial Supplies
Spécifications
- Description:Air Gun with Volume Control
- Pressure:120 psig
- Weight:0.7 lbs.
- Inlet Size:¼" FNPT
- Supplier No.:335-74SK
Spécifications
A propos de cet article
This air gun body casting with aluminum.
Delivery: Venturi nozzle delivers 100% more thrust compared to conventional air guns.
Customers Who Bought This Also Bought