ContactUsLinkComponent
Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
# de catalogue 95059-818
Fournisseur:  Thermo Scientific
Numéro CAS:  
Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
# de catalogue 95059-818
Fournisseur:  Thermo Scientific
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Fibrinogen Assay Kit
  • Cat. no.:
    95059-818
  • Supplier No.:
    100600
  • No. of Tests:
    100
Spécifications
A propos de cet article
These assays are used to screen and quantify plasma fibrinogen based on the Clauss method
They are useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot, and detect acquired and congenital hemorrhagic disorders. The tests have a reconstituted stability of 8 hours when stored at 2 to 8°C (36 to 46°F). Test components are also available separately.
Customers Who Bought This Also Bought