ContactUsLinkComponent
Protocol™ Parasitology Concentrate Systems, Thermo Scientific
Protocol™ Parasitology Concentrate Systems, Thermo Scientific
# de catalogue 95059-770
Fournisseur:  Thermo Scientific
Protocol™ Parasitology Concentrate Systems, Thermo Scientific
# de catalogue 95059-770
Fournisseur:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    50 mL Concentrate System
  • Cat. no.:
    95059-770
  • Supplier No.:
    23-005-50
Spécifications
A propos de cet article
These complete systems are for concentrating and recovering helminth eggs, larvae, and protozoan cysts from stool specimens
These complete systems are for concentrating and recovering helminth eggs, larvae, and protozoan cysts from stool specimens
Proper use optimizes isolation and identification while offering an easy-to-use processing system that minimizes specimen handling. Specimens collected and transported in 10% formalin, SAF, or empty vials may be used with these systems
Ordering information: The 50mL concentration system contains 120 filtering devices, 120 conical centrifuge tubes and screw caps, and 30mL surfactant. The 15mL concentrate system contains 50 filtering devices, 50 disposable 15mL centrifuge tubes and caps, Reagent A (surfactant), and Reagent B (ethyl acetate).
Customers Who Bought This Also Bought