Scalpel Handles, Integra™ Miltex®
Supplier: Miltex
Certificates
About this item
Handles feature mm and cm graduations
Stainless steel. Extra fine.
Ordering information: Scalpel blades are sold separately; contact you Avantor representative for more information.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.