VWR® Stackable Petri Dish Incubation Tray
# de catalogue 60914-210
Fournisseur: VWR International
Spécifications
- Cat. no.:60914-210
- W×D×H:25.1 x 23.7 x 3.5 cm (97/8 x 95/16 x 13/8")
Spécifications
A propos de cet article
Stackable Petri dish Incubation Trays increase overall capacity of an incubator by holding five 100mm diameter or fifteen 60mm diameter Petri dishes.
- Holes in tray bottom facilitate air flow
- Polypropylene construction can be disinfected with 70% Ethanol or common laboratory disinfectants
- Not recommended for use above 100°C (212°F)
- Autoclaving may warp
Each tray includes two large label areas for critical sample identification by marker or label.
Customers Who Bought This Also Bought