SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Bags feature a pre-printed sterilization indicator patch that darkens when autoclaved.
- Pre-printed sterilization indicator patch changes color with autoclaving
- Prominently displayed biohazard warning symbol
High temperature steam autoclavable to 135 °C (275 °F).
Standard bags are made from virgin-grade polypropylene that meets or exceeds the Dart Impact Resistance Test requirement of 165 g per ASTM D1709-98 and the ASTM 1922 Tear Resistance Tests. Super Biohazard Disposal Bags are made from a blended polymer that offers maximum protection against dangerous tears and leaks.
Labeling displays standard biohazard warning symbol and precautionary protocols in English, French, German, and Spanish.
Customers Who Bought This Also Bought