TearTech® 25 Holes Foam Rack, Therapak®
# de catalogue 89171-120
Fournisseur: Therapak, LLC
Spécifications
- Description:Foam rack
- Cat. no.:89171-120
- Supplier No.:68825
Spécifications
A propos de cet article
When combined with shipping boxes, specimen bags or insulated shippers, this foam tube rack provides cushioning and protection for 13x75mm tubes.
When combined with shipping boxes, specimen bags or insulated shippers, this foam tube rack provides cushioning and protection for 13x75mm tubes.
The unique die-cut construction allows for tearing away variable-sized grids to customize transport load. Overall foam block size is 150x150x100mm
Customers Who Bought This Also Bought