A propos de cet article
These versatile double mailers provide a safe environment for the transport of various laboratory specimens.
These versatile double mailers provide a safe environment for the transport of various laboratory specimens.
Each mailer comes with a 95kPa pressure tested inner receptacle, an outer fiberboard container and secure, locking Twist-n-Pull™ end caps.