About this item
Fabric boxes feature 0.64cm (1/4”) foam insulation, zipper closures, mesh envelopes for refrigerant packs, and adjustable straps for easier handling
Optional zipper locks include two matching keys. Optional security strap is reinforced with a steel cord