About this item
Kits are sealed with a peel-off foil-top lid that secures the contents.
Both kits contain an 8oz. beaker-style collection cup with a pour-spot and temperature strip, and a transport bag with absorbent material. The single-specimen kit contains a 45mL tamper-evident flip-top transport vial. The split specimen kit contains two vials.