ContactUsLinkComponent
Urine Cytology/FISH Testing Collection and Transportation Kit, Therapak®
Urine Cytology/FISH Testing Collection and Transportation Kit, Therapak®
# de catalogue 89171-030
Fournisseur:  Therapak, LLC
URINE CYTOLOGY COLL KIT 30/CS
Urine Cytology/FISH Testing Collection and Transportation Kit, Therapak®
# de catalogue 89171-030
Fournisseur:  Therapak, LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Collection and Transportation Kit
  • Cat. no.:
    89171-030
  • Supplier No.:
    37928
Spécifications
A propos de cet article
Kit contains the components necessary for urine cytology / FISH testing
The primary collection container is pre-filled with a urine preservative solution suitable for maintaining specimen viability after the urine content is transferred into the container for transport to the laboratory. To use, place two ounces of urine and two vials of preservative in the blue-cap container. The container is protected with a foam insert inside a transport bag and shipping box
Certifications: 95kPa Certifified.
Customers Who Bought This Also Bought