Linen Carts, Lakeside®
Fournisseur: Lakeside
Certificats
A propos de cet article
Compact, maneuverable carts feature four shelves and all-welded stainless steel construction
Note: Load capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
Leg and handle bumpers protect cart, walls, and doorways from damage. Blue nylon cover protects linens from airborne dust and dirt and allows for easy accessibility to the contents of the cart.
Customers Who Bought This Also Bought