ContactUsLinkComponent
Flow Cytometry/Molecular Pathology Collection and Transport Kit, Therapak®
Flow Cytometry/Molecular Pathology Collection and Transport Kit, Therapak®
# de catalogue 89171-032
Fournisseur:  Therapak, LLC
FLOW CYTOMETRY TRANS KIT 30/CS
Flow Cytometry/Molecular Pathology Collection and Transport Kit, Therapak®
# de catalogue 89171-032
Fournisseur:  Therapak, LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Transport Kit
  • Cat. no.:
    89171-032
  • Supplier No.:
    37939
Spécifications
A propos de cet article
Transport kit features all the necessary collection tools for pathology testing
To ensure safe transport, all supplies are placed into appropriate cushioning material with an absorbent pad and a secondary leak-resistant bag. The kit can be placed inside a carrier shipping envelope with attached air waybill. Kit includes two pre-filled formalin containers, slide mailer with five 75x25mm (3x1”) single frosted-end slides, one 6mL EDTA tube, and two 4mL sodium heparin tubes
Customers Who Bought This Also Bought