Albumin-Glycerol Fixative, Azer Scientific
Fournisseur: VWR International
Certificats
A propos de cet article
Fixative is used to adhere tissue specimens to glass slides in the histology laboratory.
- Adhere tissue specimens to glass slides
- Not intended for use in flotation water baths
Fixative is not intended for use in flotation water baths.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.