ContactUsLinkComponent
Dark-Line Etched Hemacytometer, Hausser Scientific
Dark-Line Etched Hemacytometer, Hausser Scientific
# de catalogue 72350-008
Fournisseur:  Hausser
Dark-Line Etched Hemacytometer, Hausser Scientific
# de catalogue 72350-008
Fournisseur:  Hausser
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Etched Hemacytometer, Complete Set
  • Cat. no.:
    72350-008
  • Supplier No.:
    3520
Spécifications
A propos de cet article
The Dark-Line etched hemacytometer is designed with an H-shaped moat that forms two counting areas, or plateaus
Contact between the flat, polished cover glass surface and the cover glass supports produces an exact volume of fluid over the counting area. The one-piece construction ensures long term durability and accurate measuring results.
Each plateau features enhanced Neubauer rulings. The loading side of each plateau features a "V" slash to facilitate filling and reduce the threat of overflow. The ruled surface rests 0.1 mm below the cover glass, limiting the volume of blood or fluid to 0.1 mm2 per square, and 0.00025 mm2 over each of the 400 squares.
BodyTextAccessories: Ideal for use in a variety of educational or research measuring and counting applications.
Ordering information: The etched hemacytometer complete set includes counting chamber and two cover slips.
Customers Who Bought This Also Bought